Learn More
Grainger Reclosable Fastener Loop 30 ft.
Supplier:  Grainger 16U573
Encompass_Preferred
The 3M Hook and Loop Fastener Roll is a 1" wide roll of hook and loop fastener tape with an adhesive backing. It has a peel adhesion of 78 oz/in and a dynamic shear strength of 25 lbs/in2. The tape is made of nylon with a pressure sensitive acrylic adhesive and has a temperature resistance range of -20°F to 200°F. This versatile fastener can bond two materials together and features hooks on one side and loops on the other for a secure grip. With 25 yards per roll, it has a wide variety of uses for mounting, attaching, and fastening in industrial, commercial, and residential applications.
Catalog No. 50-276-5800