Learn More
Grainger Reclosable Fastener Low 1 x 1 In PK100
Supplier:  Grainger 16U705
Encompass_Preferred
The 3M Dual Lock Hook and Loop Fastener Square is a reclosable fastener that creates a strong, secure hold. It features a mushroom-shaped stem with a wide top that interlocks with the opposing side for a strong bond. The fastener is made from nylon and comes in black, white or clear colors. It has an operating temperature range of -30°F to 160°F and can be cut to any shape or size. This versatile fastener can bond two rigid surfaces together or act as a reusable fastener on fabrics, plastics, metals and more. With high shear and tensile strength, it is a heavy duty option for demanding applications.
Catalog No. 50-276-6838