Learn More
Abcam Recombinant E. coli Disulfide-bond isomerase protein, 100UG
Supplier:  Abcam AB229511100UG
Encompass_Preferred
Recombinant E. coli Disulfide-bond isomerase protein is a Escherichia coli K-12 Full Length protein, in the 21 to 236 aa range, expressed in Escherichia coli, with >95% purity and suitable for SDS-PAGE, MS.
Protein/ Peptide/ Enzyme Synonyms: xprA, b2893, JW2861, dsbC, Thiol:disulfide interchange protein DsbC
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2853836