Learn More
Cell Biolabs Inc Recombinant EGFP Protein, 100 µg
Supplier:  Cell Biolabs Inc STA201
Encompass_Preferred
Expressed and purified from E. coli as a recombinant protein, greater than 90% by SDS-PAGE, Cell Biolabs’ rEGFP has an approximate MW of 29 kDa, containing a 6xHis-tag at the C-terminus, and an excitation/emission spectra of 488/507 nm. Fluorescence activity was determined by Cell Biolabs’ GFP Fluorometric Quantitation Kit #AKR-120. Provided as 100 µg/vial at 1.0 mg/mL in 1X PBS.
Catalog No. NC0567714