Learn More
Abcam Recombinant Human GDF8 / Myostatin protein (His tag), 100UG
Supplier:  Abcam AB238360100UG
Encompass_Preferred
Recombinant Human GDF8 / Myostatin protein (His tag) is a Human Full Length protein, in the 267 to 375 aa range, expressed in Escherichia coli, with >90% purity and suitable for SDS-PAGE.
Protein/ Peptide/ Enzyme Synonyms: GDF8, MSTN, Growth/differentiation factor 8, GDF-8, Myostatin
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1786846