Learn More
Abcam Recombinant human BRAF protein, 10UG
Supplier:  Abcam AB20141210UG
Encompass_Preferred
Recombinant human BRAF protein is a Human Fragment protein, in the 416 to 766 aa range, expressed in Baculovirus infected Sf9, with >80% purity and suitable for SDS-PAGE, FuncS.
Protein/ Peptide/ Enzyme Synonyms: BRAF1, RAFB1, BRAF, Serine/threonine-protein kinase B-raf, Proto-oncogene B-Raf, p94, v-Raf murine sarcoma viral oncogene homolog B1
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1955445