Learn More
Abcam Recombinant human IL-2 protein (Fc Chimera Active), 50UG
Supplier:  Abcam AB21501950UG
Encompass_Preferred
Recombinant human IL-2 protein (Fc Chimera Active) is a Human Full Length protein, in the 21 to 153 aa range, expressed in NS1, with >=98% purity, < 0.06 EU/µg endotoxin level and suitable for SDS-PAGE, FuncS.
Protein/ Peptide/ Enzyme Synonyms: Interleukin-2, IL-2, T-cell growth factor, TCGF, IL2
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1878277