Learn More
Abcam Recombinant Human Serum Albumin protein (His tag), 500UG
Supplier:  Abcam AB217817500UG
Encompass_Preferred
Recombinant Human Serum Albumin protein (His tag) is a Human Full Length protein, in the 25 to 609 aa range with >95% purity, < 1 EU/µg endotoxin level and suitable for SDS-PAGE and Functional studies. The predicted molecular weight of ab217817 protein is 67.3 kDa. - Save time and ensure accurate results - use our Serum Albumin protein as a control - Optimal protein bioactivity, stability and reproducibility
Protein/ Peptide/ Enzyme Synonyms: GIG20, GIG42, PRO0903, PRO1708, PRO2044, PRO2619, PRO2675, UNQ696/PRO1341, ALB, Albumin
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1469881