Learn More
Abcam Recombinant mouse Jagged1 protein (Fc Chimera Active), 10UG
Supplier:  Abcam AB10934610UG
Encompass_Preferred
Recombinant mouse Jagged1 protein (Fc Chimera Active) is a Mouse Fragment protein, in the 1 to 1067 aa range, expressed in HEK 293, with >90% purity, < 0.01 EU/µg endotoxin level and suitable for SDS-PAGE, Inhib, FuncS.
Protein/ Peptide/ Enzyme Synonyms: CD339, Protein jagged-1, Jagged1, Jag1
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1350486