Learn More
KACTUSBIO INC Recombinant Mouse CD98 Protein is expressed from HEK293 with His tag at the N-terminus.It contains Ala100-Ala526.
Supplier:  KACTUSBIO INC CD9MM198100UG
Encompass
Mouse CD98 Protein (CD9-MM198). Endotoxin is less than 1 EU per μg by the LAL method. The purity of this protein is determined to be > 95% by Bis-Tris PAGE and > 95% by HPLC.
Catalog No. 50-004-65219