Learn More
Abcam Recombinant Rhesus monkey IL-2 protein (Tagged), 100UG
Supplier:  Abcam AB224874100UG
Encompass_Preferred
Recombinant Rhesus monkey IL-2 protein (Tagged) is a Rhesus monkey Full Length protein, in the 21 to 154 aa range, expressed in Escherichia coli, with >90% purity and suitable for SDS-PAGE.
Protein/ Peptide/ Enzyme Synonyms: Interleukin-2, IL-2, T-cell growth factor, TCGF, IL2
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1638285