Learn More
Takara Bio Recombinant RNase Inhibitor | 25,000 Units
SDP
Supplier:  Takara Bio 2313B
Encompass
Recombinant RNase Inhibitor forms a 1:1 complex with RNase A and inhibits RNase activity. This reaction is reversible, and ribonuclease activity can be recovered by dissociating the complex with urea or sulfhydryl reagent. This reaction irreversibly inactivates the inhibitor. Recombinant RNase Inhibitor can be added directly to reaction mixtures containing RNA. In addition, because it is a protein, it differs from other competitive inhibitors (nucleotides and inorganic phosphates) by being able to be easily removed from the reaction system using phenol extraction.
Catalog No. NC1471728