Learn More
Abcam Recombinant Human beta Catenin protein (Tagged), 20UG
Supplier:  Abcam AB6317520UG
Encompass_Preferred
Recombinant Human beta Catenin protein (Tagged) is a Human Full Length protein, in the 1 to 781 aa range, expressed in Baculovirus infected Sf9, with >70% purity and suitable for SDS-PAGE, ELISA, WB.
Protein/ Peptide/ Enzyme Synonyms: CTNNB, OK/SW-cl.35, PRO2286, CTNNB1, Catenin beta-1, Beta-catenin
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1844473