Learn More
Bio X Cell Bio X Cell RecombiMAb Anti-Mouse PD-1 (CD279) (D265A), 5 mg vial
Supplier: Bio X Cell CP151R005MG
Bio X Cell RecombiMAb anti-mouse PD-1 (CD279) (D265A) is a recombinant monoclonal antibody designed for in vivo blockade of PD-1 signaling in mouse models. It combines the RMP1-14 variable domains with a mouse IgG2a Fc bearing a D265A mutation to reduce Fcγ receptor binding, is purified from CHO cell supernatant, and is formulated in PBS, pH 7.0.
- Recombinant monoclonal antibody targeting mouse PD-1 (CD279).
- Clone RMP1-14 with mouse IgG2a Fc and a D265A mutation.
- Purified from CHO cell supernatant and formulated in PBS, pH 7.0.
- Purity ≥95% and endotoxin ≤0.5 EU/mg.
- Contains no stabilizers or preservatives.
- Intended for in vivo blocking of PD-1/PD-L signaling.
- Available in multiple sizes, including a 5 mg vial.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.