Learn More
Bio X Cell RecombiMAb Anti-Mouse OX40 (CD134) Antibody, Clone OX86-CP017, 1 mg, Mouse IgG2a
Supplier: Bio X Cell CP0171MG
RecombiMAb anti-mouse OX40 (CD134) is a chimeric recombinant antibody (clone OX86) engineered to activate mouse OX40 in in vivo and in vitro studies. The product uses mouse IgG2a constant regions with variable domains matching the original OX-86 clone, is supplied in PBS (pH 7.0), and is provided at 1 mg.
- Clone OX86 recombinant antibody.
- Isotype mouse IgG2a, kappa.
- Reactivity mouse.
- Target OX40 (CD134).
- Applications in vivo activation, in vitro activation, and western blot.
- Formulated in PBS, pH 7.0; no added stabilizers or preservatives.
- Purity ≥95%; aggregation <5%; endotoxin ≤0.5 EU/mg.
- Sterile filtered (0.2 μm) and stored at 4°C; do not freeze.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.