Learn More
Bio X Cell RecombiMAb Human IgG1 Isotype Control Antibody (anti-hen egg lysozyme), 1 MG, 1 MG
Supplier: Bio X Cell CP174R001MG
RecombiMAb human IgG1 isotype control antibody that recognizes hen egg lysozyme and shows minimal or no specific binding to human samples. The antibody is formulated in PBS (pH 7.0), supplied in multiple vial sizes including 1 mg, and should be stored at 4C without freezing.
- Isotype control for human IgG1.
- Binds hen egg lysozyme with minimal reactivity to human samples.
- Formulated in PBS, pH 7.0.
- Available in 1 mg and larger sizes for in vivo or in vitro use.
- Store at 4C; do not freeze.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.