Learn More
Abcam Recombinant Human CTGF protein, 50UG
Supplier:  Abcam AB28345750UG
Encompass_Preferred
Recombinant Human CTGF protein is a Human Full Length protein, in the 27 to 349 aa range, expressed in HEK 293, with >=95% purity and suitable for SDS-PAGE.
Protein/ Peptide/ Enzyme Synonyms: CTGF, HCS24, IGFBP8, CCN2, CCN family member 2, Cellular communication network factor 2, Connective tissue growth factor, Hypertrophic chondrocyte-specific protein 24, Insulin-like growth factor-binding protein 8, IBP-8, IGF-binding protein 8, IGFBP-8
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2059560