Learn More
Sino Biological Recombinant Anti-CD96 Antibody, Rabbit Monoclonal 100 µL
Supplier:  Sino Biological 11202R148100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD96 (rh CD96; Catalog#11202-H08H; P40200-2; Met1-Met503).
Catalog No. 50-007-49490