Learn More
Abcam Anti-cleaved N-terminal DFNA5 / GSDME antibody [EPR20866-160] - BSA and Azide free
Supplier:  Abcam AB256122100UG
Encompass_Preferred
Rabbit monoclonal [EPR20866-160] to cleaved N-terminal DFNA5 / GSDME - BSA and Azide free
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2608486