Learn More
Sino Biological Recombinant Anti-EPCR Antibody (PE), Rabbit Monoclonal 25 Tests
Supplier:  Sino Biological 50874R016P25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Epcr/PROCR (rM Epcr/PROCR; Catalog#50874-M08H; Q64695; Met1-Ser214) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-007-85383