Learn More
Sino Biological Recombinant Anti-NCAM-1/CD56 Antibody, Rabbit Monoclonal 100 µL
Supplier: Sino Biological 10673R266100
This antibody was obtained from a rabbit immunized with purified, recombinant Human NCAM-1/CD56 (rh NCAM-1/CD56; Catalog#10673-H08H; NP_001070150.1; Met1-Pro603).