Learn More
ILEX LIFE SCIENCES LLC Recombinant CTGF Human Expressed in E coli 5 ug 5 UG 1/EA
SDP
Supplier:  ILEX LIFE SCIENCES LLC A35410005
Encompass_Preferred
Ilex Life Sciences Connective Tissue Growth Factor (CTGF) Human E coli Recombinant Protein 5 g DESCRIPTION Our bioactive CTGF Human E coli Recombinant Protein is a single non-glycosylated polypeptide chain containing 98 amino acids It has a molecular mass of 11 2 kDa SOURCE Expressed in E coli FORMULATION Sterile-filtered white lyophilized powder Lyophilized from a sterile filtered aqueous solution containing 0 1% Trifluoroacetic Acid (TFA) PURITY 90% (SDS-PAGE) BIOACTIVITY Determined by the dose-dependent stimulation of the proliferation of HUVEC cells The expected ED50 for this effect is 1-2 g/ml corresponding to a specific activity of 500-1000 units/mg OTHER NAMES CCN family member 2 CCN2 Hypertrophic chondrocyte-specific protein 24 HCS24 Insulin-like growth factor-binding protein 8 IGF-binding protein 8 IGFBP-8 IBP-8 UniProtKB P29279 USAGE For laboratory research use only Not for use in diagnostic or therapeutic procedures
Catalog No. 50-005-80403