Learn More
Active Motif Recombinant EGFR protein (672-1210, L858R) protein
SDP
Supplier:  Active Motif 81200
Encompass
EGFR (Epidermal Growth Factor Receptor), also known as ERBB, mENA, ERBB1 and HER1, is the receptor for members of the EGF family and is a transmembrane glycoprotein that has tyrosine kinase activity. Receptor tyrosine kinase binding ligands of the EGF family and activating several signaling cascades to convert extracellular cues into appropriate cellular responses. Known ligands include EGF, TGFA/TGF-alpha, AREG, epigen/EPGN, BTC/betacellulin, epiregulin/EREG and HBEGF/heparin-binding EGF. Ligand binding triggers receptor homo and/or heterodimerization and autophosphorylation on key cytoplasmic residues. The phosphorylated receptor recruits adapter proteins like GRB2 which in turn activates complex downstream signaling cascades, including at least 4 major downstream signaling cascades: the RAS-RAF-MEK-ERK, PI3 kinase-AKT, PLCgamma-PKC and STATs modules, leading to cell proliferation. […] For more information, please visit: https://www.activemotif.com/catalog/details/81200.
Catalog No. NC1852151