Learn More
ILEX LIFE SCIENCES LLC Recombinant Erythropoietin Alpha Human Expressed in CHO cells 1 mg 1 MG 1/EA
SDP
Supplier:  ILEX LIFE SCIENCES LLC A32011000
Encompass_Preferred
Ilex Life Sciences Erythropoietin Alpha (EPO) Human CHO Recombinant Protein 1 mg DESCRIPTION Erythropoietin Alpha (EPO) Human CHO Recombinant Protein is a single polypeptide chain that contains 166 amino acids and has a predicted molecular mass of 21 000 Dalton and apparent glycosylated molecular mass of 36-40 kDa SOURCE Expressed in Chinese hamster ovary (CHO) cells FORMULATION Sterile-filtered white lyophilized (freeze-dried) powder Each mg of lyophilized powder contains 0 59 mg sodium citrate 0 58 mg sodium chloride and 0 006 mg citric acid PURITY 98 0% (RP-HPLC and SDS-PAGE) BIOACTIVITY The Specific Activity was measured by normocythemic mice bioassay and was found to be 150 000 IU/mg (NOTE may vary with each lot number Please refer to the datasheet enclosed with your product) OTHER NAMES Erythropoietin-alpha EPOa EPO-alpha EPO-a USAGE This product is intended for Laboratory Research Use Only Not for use in diagnostic or therapeutic procedures
Catalog No. 50-005-80406