Learn More
ACROBiosystems ACROBiosystems SAFENSURE Recombinant Factor C Endotoxin Detection Kit, 96 tests (RES-A056-96tests)
Supplier:  ACROBiosystems RESA05696TESTS
Encompass_Preferred
Fluorescence-based recombinant Factor C assay for quantitative detection of bacterial endotoxin in biological samples and process materials. The kit uses recombinant Factor C to cleave a fluorogenic substrate; fluorescence is measured after incubation and correlates with endotoxin concentration. Intended for research use in process control and product testing.
  • Quantitative detection range 0.005-5 EU/mL.
  • Endpoint fluorescent microplate assay compatible with 96-well readers.
  • Measured at ex/em = 380/440 nm after 37 °C incubation for 1 hour.
  • Includes endotoxin standard, recombinant Factor C reagent, fluorogenic substrate, endotoxin-free water, and a 96-well endotoxin-free plate.
  • Suitable for raw material, water, intermediate, and final product testing in research workflows.
  • Store unopened kit at 2-8 °C and do not use reagents beyond expiration.
Catalog No. NC3835967