Learn More
ILEX LIFE SCIENCES LLC Recombinant FGF10 Human Expressed in E coli 1 mg 1 MG 1/EA
SDP
Supplier:  ILEX LIFE SCIENCES LLC A33031000
Encompass_Preferred
Ilex Life Sciences FGF-10 (KGF-2) Human E coli Recombinant Protein 1 mg DESCRIPTION Our bioactive FGF-10 (KGF-2) Human E coli Recombinant Protein is a single non-glycosylated polypeptide chain that contains 170 amino acids (a a 40-208) and has a molecular mass of 19300 Dalton The FGF-10 protein is recombinantly produced in E coli SOURCE Expressed in E coli FORMULATION Sterile-filtered white lyophilized (freeze-dried) powder Lyophilized from a 0 2 m filtered solution (1 mg/ml) in PBS pH 7 4 PURITY 96 0% (RP-HPLC and SDS-PAGE) BIOACTIVITY The ED50 calculated by the dose-dependent stimulation of FGF receptors by BaF3 indicator cells (measured by 3H-thymidine uptake) is 0 5 ng/ml corresponding to a specific activity of 2x10 6 units/mg OTHER NAMES Human KGF-2 Keratinocyte growth factor 2 Human FGF-10 Human FGF10 Fibroblast growth factor 10 USAGE Intended for Laboratory Research Use Only Not for use in diagnostic or therapeutic procedures
Catalog No. 50-005-80423