Learn More
ILEX LIFE SCIENCES LLC Recombinant FGF10 Mouse Expressed in E coli 5 ug 5 UG 1/EA
SDP
Supplier:  ILEX LIFE SCIENCES LLC A31260005
Encompass_Preferred
Ilex Life Sciences FGF-10 (KGF-2) Mouse E coli Recombinant Protein 5 g DESCRIPTION FGF-10 Mouse E coli Recombinant Protein is a single non-glycosylated polypeptide chain that contains 173 amino acids and has a 19 5 kDa molecular mass SOURCE Expressed in E coli FORMULATION Sterile-filtered white lyophilized (freeze-dried) powder The protein was lyophilized from a 0 2 m filtered concentrated solution in 1 PBS pH 7 4 containing 5% trehalose PURITY 97 0% as determined by RP-HPLC and SDS-PAGE BIOACTIVITY Fully biologically active when compared to standard The ED50 as determined by the dose-dependent stimulation of thymidine uptake by BaF3 cells expressing FGF receptors is 0 5 ng/ml OTHER NAMES Mouse KGF-2 Murine KGF2 Keratinocyte growth factor 2 Mouse FGF-10 Mouse FGF10 Fibroblast growth factor 10 USAGE This product is intended for Laboratory Research Use Only Not for use in diagnostic or therapeutic procedures
Catalog No. 50-005-80366