Learn More
Abcam Recombinant Hepatitis B virus Protein X (His tag), 100UG
Supplier:  Abcam AB236464100UG
Encompass_Preferred
Recombinant Hepatitis B virus Protein X (His tag) is a Hepatitis B virus adr4 Fragment protein, in the 2 to 154 aa range, expressed in Escherichia coli, with >90% purity and < 1 EU/µg endotoxin level.
Protein/ Peptide/ Enzyme Synonyms: Protein X, HBx, Peptide X, pX, X
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-6412