Learn More
Abcam Recombinant Human SOX9 protein, 10UG
Supplier:  Abcam AB13191110UG
Encompass_Preferred
Recombinant Human SOX9 protein is a Human Full Length protein, in the 1 to 509 aa range, expressed in Wheat germ and suitable for SDS-PAGE, ELISA, WB.
Protein/ Peptide/ Enzyme Synonyms: Transcription factor SOX-9, SOX9
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2209008