Learn More
Abcam Recombinant Human Tau441 protein, 50UG
Supplier:  Abcam AB8470050UG
Encompass_Preferred
Recombinant Human Tau441 protein is a Human Full Length protein, in the 1 to 441 aa range, expressed in Escherichia coli, with >90% purity and suitable for SDS-PAGE, WB.
Protein/ Peptide/ Enzyme Synonyms: UNQ3115/PRO10196, FGF21, Fibroblast growth factor 21, FGF-21
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1994375