Learn More
Abcam Recombinant Human ACTR8 protein, 10UG
Supplier:  Abcam AB17285110UG
Encompass_Preferred
Recombinant Human ACTR8 protein is a Human Full Length protein, in the 1 to 329 aa range, expressed in Escherichia coli, with >95% purity, < 1 EU/µg endotoxin level and suitable for SDS-PAGE, HPLC.
Protein/ Peptide/ Enzyme Synonyms: ARP8, INO80N, ACTR8, Actin-related protein 8, hArp8, INO80 complex subunit N
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-8260