Learn More
Creative Biomart Trafermin, Recombinant human basic fibroblast growth factor Protein
Supplier:  Creative Biomart THP0292
Encompass_Preferred
Trafermin (brand name Fiblast), also known as recombinant human basic fibroblast growth factor (rhbFGF), is a recombinant form of human basic fibroblast growth factor (bFGF) which is marketed in Japan as a topical spray for the treatment of skin ulcers. It is also currently in preregistration for the treatment of periodontitis. As a recombinant form of bFGF, trafermin is a potent agonist of the FGFR1, FGFR2, FGFR3, and FGFR4. The drug has been marketed in Japan since June 2001.
Catalog No. 50-208-4342