Learn More
Medchemexpress LLC Recombinant human CD131 protein (HEK293, His) | >95.0% | 500 UG

Supplier: Medchemexpress LLC HYP75608500UG
Recombinant human CD131 extracellular domain (W17-W443) with a C-terminal His tag, expressed in HEK293 cells and supplied in sterile PBS for research applications such as binding assays and antibody characterization.
- Extracellular domain covering W17-W443.
- C-terminal His tag for purification and detection.
- Greater than 95.0% purity by reducing SDS-PAGE.
- Supplied in 0.22 μm filtered PBS, pH 7.4.
- Stable at -80°C for long-term storage; avoid repeated freeze-thaw cycles.
- Available in multiple research sizes, including 500 μg.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.