Learn More
Creative Biomart Recombinant Human CD19 protein, His-tag, GMP Grade
Supplier:  Creative Biomart CD19307THP
Encompass_Preferred
Recombinant Human CD19 protein GMP Grade(NP_001171569)(271 aa) was expressed in HEK293 cells with a C-terminal His Tag.
Catalog No. 50-208-5293