Learn More
Abcam Recombinant human CXCL10/IP10 protein (Active), 100UG
Supplier:  Abcam AB280332100UG
Encompass_Preferred
Recombinant human CXCL10/IP10 protein (Active) is a Human Full Length protein, in the 22 to 98 aa range with >=95% purity, <= 0.005 EU/µg endotoxin level and suitable for SDS-PAGE, Functional studies, mass spectrometry and HPLC. The predicted molecular weight of ab280332 protein is 8.7 kDa. - Save time and ensure accurate results - use our CXCL10/IP10 protein as a control - Optimal protein bioactivity, stability and reproducibility - Available in different sizes to fit your experimental needs
Protein/ Peptide/ Enzyme Synonyms: INP10, SCYB10, CXCL10, C-X-C motif chemokine 10, 10 kDa interferon gamma-induced protein, Small-inducible cytokine B10, Gamma-IP10, IP-10
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2070141