Learn More
Abcam Recombinant Human Cytochrome C protein, 10UG
Supplier:  Abcam AB17281710UG
Encompass_Preferred
Recombinant Human Cytochrome C protein is a Human Full Length protein, in the 2 to 105 aa range, expressed in Escherichia coli, with >95% purity, < 1 EU/µg endotoxin level and suitable for SDS-PAGE, HPLC.
Protein/ Peptide/ Enzyme Synonyms: CYC, CYCS, Cytochrome c
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1827514