Learn More
Creative Biomart Nepidermin, Recombinant human epidermal growth factor (rhEGF) Protein
Supplier:  Creative Biomart THP0303
Encompass_Preferred
Nepidermin (brand name Easyef), also known as recombinant human epidermal growth factor (rhEGF), is a recombinant form of human epidermal growth factor (EGF) and a cicatrizant (a drug that promotes wound healing through formation of scar tissue). It was developed by Daewoong Pharmaceutical. As a recombinant form of EGF, nepidermin is an agonist of the epidermal growth factor receptor (EGFR), and is the first EGFR agonist to be marketed.
Catalog No. 50-208-4353