Learn More
Abcam Recombinant human FGF2 protein (Active), 50UG
Supplier:  Abcam AB28546250UG
Encompass_Preferred
Recombinant human FGF2 protein (Active) is a Human Full Length protein, expressed in Escherichia coli, with >95% purity, < 1 EU/µg endotoxin level and suitable for FuncS.
Protein/ Peptide/ Enzyme Synonyms: FGFB, FGF2, Fibroblast growth factor 2, FGF-2, Basic fibroblast growth factor, Heparin-binding growth factor 2, bFGF, HBGF-2
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-220-5543