Learn More
Creative Biomart Recombinant Human FGF7 protein, Fc-tag, GMP Grade
Supplier:  Creative Biomart FGF7411THP
Encompass_Preferred
Recombinant Human FGF7 protein GMP Grade(NP_002000.1)(292 aa) was expressed in HEK293 cells with a C-terminal Fc Tag.
Catalog No. 50-208-5272