Learn More
Abcam Recombinant human FKBP12 protein, 500UG
Supplier:  Abcam AB85840500UG
Encompass_Preferred
Recombinant human FKBP12 protein is a Human Full Length protein, expressed in Escherichia coli, with >95% purity, < 1 EU/µg endotoxin level and suitable for SDS-PAGE, FuncS.
Protein/ Peptide/ Enzyme Synonyms: FKBP1, FKBP12, FKBP1A, Peptidyl-prolyl cis-trans isomerase FKBP1A, PPIase FKBP1A, 12 kDa FK506-binding protein, Calstabin-1, FK506-binding protein 1A, Immunophilin FKBP12, Rotamase, 12 kDa FKBP, FKBP-12, FKBP-1A
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2231611