Learn More
Abcam Recombinant Human GLP-1R protein (His tag), 100UG
Supplier:  Abcam AB239555100UG
Encompass_Preferred
Recombinant Human GLP-1R protein (His tag) is a Human Fragment protein, in the 24 to 145 aa range, expressed in Yeast, with >90% purity and suitable for SDS-PAGE.
Protein/ Peptide/ Enzyme Synonyms: Glucagon-like peptide 1 receptor, GLP-1 receptor, GLP-1-R, GLP-1R, GLP1R
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2227276