Learn More
Abcam Recombinant Human GLP-1R protein, 100UG
Supplier:  Abcam AB130280100UG
Encompass_Preferred
Recombinant Human GLP-1R protein is a Human Fragment protein, in the 24 to 145 aa range, expressed in Escherichia coli, with >90% purity, < 1 EU/µg endotoxin level and suitable for SDS-PAGE, WB.
Protein/ Peptide/ Enzyme Synonyms: Glucagon-like peptide 1 receptor, GLP-1 receptor, GLP-1-R, GLP-1R, GLP1R
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2227253