Learn More
Medchemexpress LLC Recombinant human Granzyme A protein | 143180-73-8 | >95.0% | 5 UG

Supplier: Medchemexpress LLC HYP763775UG
Recombinant human Granzyme A (GZMA) expressed in HEK293 cells with a C-terminal His tag. Supplied as a lyophilized, purified protein corresponding to amino acids E27-V262 and intended for in vitro biochemical and enzymatic assays; the proenzyme requires activation for full enzymatic activity.
- Expressed in HEK293 cells
- C-terminal His tag for purification and detection
- Purity greater than 95% by reducing SDS-PAGE
- Molecular weight approximately 27-33 kDa
- Supplied lyophilized, 5 μg vial
- Specific activity greater than 5,000 pmol/min/μg (requires activation)
- Recommend freezing aliquots at -20°C or -80°C for extended storage
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.