Learn More
Abcam Recombinant Human Histone H2A (biotinylated ) protein, 50UG
Supplier:  Abcam AB20028650UG
Encompass_Preferred
Recombinant Human Histone H2A (biotinylated ) protein is a Human Full Length protein, in the 2 to 130 aa range, expressed in Escherichia coli, with >56% purity and suitable for SDS-PAGE.
Protein/ Peptide/ Enzyme Synonyms: H2AW, HIST3H2A, H2AC25, Histone H2A type 3, H2A-clustered histone 25
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1703118