Learn More
Abcam Recombinant Human HLA G protein (His tag), 10UG
Supplier:  Abcam AB22566010UG
Encompass_Preferred
Recombinant Human HLA G protein (His tag) is a Human Full Length protein, in the 25 to 338 aa range, expressed in Mammalian, with >90% purity and suitable for SDS-PAGE, MS.
Protein/ Peptide/ Enzyme Synonyms: HLA-6.0, HLAG, HLA-G, HLA G antigen, MHC class I antigen G
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1673708