Learn More
Abcam Recombinant Human Hsp90 beta protein, 200UG
Supplier:  Abcam AB80353200UG
Encompass_Preferred
Recombinant Human Hsp90 beta protein is a Human Full Length protein, expressed in Escherichia coli, with >95% purity and suitable for SDS-PAGE.
Protein/ Peptide/ Enzyme Synonyms: HSP90B, HSPC2, HSPC3, HSPCB, HSP90AB1, Heat shock protein HSP 90-beta, HSP 90, Heat shock 84 kDa, Heat shock protein family C member 3, HSP 84, HSP84
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1881132