Learn More
Abcam Recombinant human Interferon alpha 1 protein (Active), 100UG
Supplier:  Abcam AB48750100UG
Encompass_Preferred
Recombinant human Interferon alpha 1 protein (Active) is a Human Full Length protein, in the 24 to 189 aa range, expressed in Escherichia coli, with >95% purity, < 1 EU/µg endotoxin level and suitable for SDS-PAGE, sELISA.
Protein/ Peptide/ Enzyme Synonyms: IFNA13, IFNA1, Interferon alpha-1/13, IFN-alpha-1/13, Interferon alpha-D, LeIF D
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1120842