Learn More
Abcam Recombinant Human L3MBTL1 protein, 100UG
Supplier:  Abcam AB216202100UG
Encompass_Preferred
Recombinant Human L3MBTL1 protein is a Human Fragment protein, in the 191 to 530 aa range, expressed in Escherichia coli, with >=88% purity and suitable for SDS-PAGE.
Protein/ Peptide/ Enzyme Synonyms: KIAA0681, L3MBT, L3MBTL, L3MBTL1, Lethal(3)malignant brain tumor-like protein 1, H-l(3)mbt, H-l(3)mbt protein, L(3)mbt-like, L(3)mbt protein homolog
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-6671