Learn More
Abcam Recombinant Human Lactate Dehydrogenase protein, 100UG
Supplier:  Abcam AB235557100UG
Encompass_Preferred
Recombinant Human Lactate Dehydrogenase protein is a Human Fragment protein, in the 5 to 323 aa range, expressed in Escherichia coli, with >85% purity and suitable for SDS-PAGE.
Protein/ Peptide/ Enzyme Synonyms: PIG19, LDHA, L-lactate dehydrogenase A chain, LDH-A, Cell proliferation-inducing gene 19 protein, LDH muscle subunit, Renal carcinoma antigen NY-REN-59, LDH-M
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-6059