Learn More
Abcam Recombinant human LIM Kinase 1 protein, 10UG
Supplier:  Abcam AB17406610UG
Encompass_Preferred
Recombinant human LIM Kinase 1 protein is a Human Fragment protein, in the 285 to 638 aa range, expressed in Baculovirus infected Sf9, with >95% purity and suitable for SDS-PAGE, WB, FuncS.
Protein/ Peptide/ Enzyme Synonyms: LIMK, LIMK1, LIM domain kinase 1, LIMK-1
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2112650